Processor Node Missing
Issue
User had Proteome Discoverer 2.1 on one computer, and that computer died, so she had to reinstall Proteome Discoverer 2.1 on a new computer. When she did, the MS2 spectrum processor node which had been on Proteome Discoverer 2.1 on the previous computer was absent from the new computer.
Environment
- MS2 spectrum processor
- Proteome Discoverer 2.1
Resolution
User went to the following website and was able to download the node from there:
http://ms.imp.ac.at/index.php?action=spectrum-processor.