EQuan MAX Plus upgrade procedure
Description:
- EQuan MAX Plus was released for DCMSLink this is not supported anymore on new Foundation version and under W10.
- EQuan MAX Plus with DCMSLink can not perform 20mL, but it is written in product specification sheet.
Environment:
- EQuan MAX Plus
- DCMSLink
- Autosampler
- SII
Attachment(s):
| File | Last Modified |
|---|---|
| EQuan MAX Plus upgrade procedure.pdf | December 31, 2021 |