Thermo Fisher Scientific
DXR Failing USP Photometric Precision
DXR Failing USP Photometric Precision
Issue
When running Valpro on the DXR Smart Raman the  USP Photometric Precision tests fail.
Environment
  • DXR SmartRaman
Resolution
Navigate to the folder C:\my documents\omnic
Delete the file "options.top"
When the Valpro test is run the next time, the "options.top" file will be recreated with new photometric precision baseline values for the instrument.
Analysis
Cause
  • Was this article helpful?