DXRxi Laser Alignment fails / no "Live Spectrum"
Issue
On DXRxi-Systems the Alignment/Calibration fails during "Laser Alignment" or is pending for a very long time (>1h). "Live Spectrum" is not working.
Environment
DXRxi
DXR2xi
DXR3xi
Resolution
Aperture assembly stick causing alignment, calibration, and data collection errors.
Refer to: DXR and DXRxi Stuck Aperture Recovery, Procedure number: DXRxi 023.
Attachment(s)
| File | Last Modified |
|---|---|
| DXRxi_023_-_Rev_00_-_DXR_and_DXRxi_Stuck_Aperture_Recovery.pdf | June 25, 2024 |