DXR first generation Spectrograph driver
Issue
Drivers from original spectrograph are no longer included in current versions of OMNIC software, which is causing the system to not communicate.
Environment
- OMNIC versions greater than 9.3
- Raman DXR
Resolution
Attached is a file with the 64bit drivers from the 9.3 update DVD. Download the attached file and extract files to a folder and update driver from device manager.
Attachment(s)
| File | Last Modified |
|---|---|
| DXR Original Spectrograph Driver 64bit.zip | March 14, 2022 |