DXRxi has no spectrograph signal and fails Alignment
Issue
DXRxi has no spectrograph signal and fails Alignment
Environment
- DXR2xi
- DXR3xi
Resolution
Check BNC cable from stage controller to BNC on back of system to make sure it is not damaged. Check cable for continuity if cable fails replace the cable part number 085-805600
Analysis
Cause