Find laser usage time on DXRxi system
Issue
The OMNICxi software does not display the laser hours
Environment
- DXRxi instruments
Resolution
The iButton on the laser stores the time. To find the laser usage time for current laser use the following steps:
- Open Map_ServiceToolApp (C:\Program Files\Thermo Scientific\OMNICxi)
- Password is (passworddate - replace date with day of month)
- Click on iButtons
- Laser Usage listed in minutes