Low Frequency Limit of Grating Message in OMNIC
The article addresses an issue with OMNIC software displaying a "Low frequency limit of grating (39.3) is outside the allowed range of 35 to 5 cm-1" message in the context of DXR3 SmartRaman and DXR3 Microscope. The primary causes include thermal stabilization required after grating installation, debris or dirt on the grating magnets, and missing locating rods or ball bearings in the spectrograph. Solutions involve allowing time for thermal stabilization, cleaning the magnets, and checking for n
Issue
OMNIC software is giving a message that say "Low frequency limit of grating (39.3) is outside the allowed range of 35 to 5 cm-1".
Environment
DXR3 SmartRaman
DXR3 Microscope
OMNIC for Dispersive Raman
Resolution
This message can be caused by the following:
- The most common cause is that the grating was recently installed and has not achieved thermal stabilization. After installing a grating allow 15 to 20 minutes for warm up and stabilization.
- Another cause is if there is some debris or dirt on the magnets of the grating or in the spectrograph. Ensure all magnets are clean and remove an debris that is found.
- The last possible cause could be if one of the locating rods on the grating or a ball bearing in the spectrograph is missing. Check that these are in place.
Attachment(s)
| File | Last Modified |
|---|---|
| DXR - Low Frequency Limit of Grating messages.png | September 13, 2025 |