OMNICxi Stops Immediately at Laser Calibration Step
Issue
When running the alignment and calibration routine in OMNICxi, the process stops as soon as it gets to step 6 for Laser Calibration
Environment
OMNICxi v2.5
DXR3xi
Resolution
Open the ServiceMapTool.exe program and click on the EEPROM item.
In the EEPROM window under the Laser Power Calibration section, use the drop down to select the laser wavelength that is failing.
Check the Power Sensor and four NDF parameter fields to ensure they have values entered.
If these fields are blank, enter the correct values or restore from the dxreeprom.dat file and then write to the board.
Analysis
Cause