Thermo Fisher Scientific
Angle of Incidence for Nicolet iN10 15X objective
Issue
What are the minimum and maximum angles of incidence of the light through the Nicolet iN10 15X objective
Environment
Nicolet iN10
Nicolet iN10MX
Resolution
The minimum angle of incidence is 20 degrees
The maximum angle of incidence is 43.5 degrees
The average angle of incidence is 31.75 degrees