Bench status report
Issue
Which FT-IR benches use the bench status report?
Environment
FT-IR
Resolution
The iS5, iS10, iS20, and iS50 Spectrometer benches use the bench status report.
Skip to main content
Which FT-IR benches use the bench status report?
FT-IR
The iS5, iS10, iS20, and iS50 Spectrometer benches use the bench status report.