Cannot write archive in database
Issue
Why do I get “Cannot write archive in database” error message?
Environment
- RESULT
- Antaris
Resolution
- Exit from RESULT software.
- Rename RESULT.mdb file located in C:\RESULT Data\Config.
- RESULT software will create a new database file next time RESULT software is run.
Cause
The error message is mostly likely due to a corrupt database file.