Diamond Phonon Explanation
Description:
Diamond crystals used during infrared analysis has natural absorbances that occur between 2600 and 1600 cm-1 in the mid-Infrared region and represent the phonon bands within the structure of the diamond crystal.
The attachment describes this phenomena.
Environment:
Nicolet Continuum
Attachment(s)
File | Last Modified |
---|---|
Diamond Phonon Explanation.pdf | December 27, 2021 |