FTIR Auto-Align
Issue
Should the FTIR be auto-aligned after changing the Smart accessory or IR detector?
Environment
FT-IR
Resolution
Auto-alignment should be performed:
- After replacing the beamsplitter;
- Periodically every couple of weeks; and
- Before a critical set of experiments.