Thermo Fisher Scientific
Nicolet iN10 Ge Tip ATR Angle of Incidence
Issue
What is the angle of incidence for the slide in Ge ATR tip used on the Nicolet iN10 microscope.
Environment
Nicolet iN10
Nicolet iN10MX
Resolution
The average angle of incidence of the slide on Ge Tip ATR for the Nicolet iN10 is 27 degrees.
Analysis
The angle of incidence may be needed to apply the Advanced ATR correction to a spectrum collected with the iN10 Ge ATR.