Summit computer low on disk space. Windows reports hard drive is full.
Issue
Summit computer low on disk space. Windows reports hard drive is full. and Paradigm will not open
Environment
- Summit Pro
- Summit
Resolution
Check C:\ProgramData\Thermo Scientific\Paradigm\DbBackup. If there are multiple backup folders located here delete them.
Check C:\ProgramData\RabbitMQ\Log. If the log files are larger then 1GB the log files can be deleted to restore drive space on the Summit SOM.
Check %temp% folder and Downloads folder and delete unneeded files if any.
After deleting files Recycle Bin should be emptied.
Attachment(s)
File | Last Modified |
---|---|
Large RabbitMQ log file.PNG | November 13, 2023 |