iS5 Failing the wavelength accuracy during Performance verification
Issue
Nicolet iS5 Failing the wavelength accuracy during Performance verification
Environment
- Nicolet iS5
Resolution
- To fix this issue run iS5 Laser Verify H2O.mac by adding it to the Omnic tool bar from C:\my documents\omnic\Param\factory\LaserCal\
- After running Macro accept new laser Frequency and rerun Performance verification.
Analysis
Cause