Updating NSS Software
Description:
Assure ALL NSS Peripherals Are Powered UP When Updating NSS Software
Environment:
- Noran System 6
- Noran System 7
- Pathfinder
RESOLUTION:
- If ANY NSS peripheral is not turned on when performing any NSS software update, various problems will occur.
- Assure all NSS peripherals are turned on when updating any NSS software.
- These include the NSS Analyzer Chassis and any WDS or EBSD controller. The NSS software install will also update software to these peripherals (if present).