Thermo Fisher Scientific
About OMNIC Paradigm software
Use OMNIC Paradigm software Operator mode to run the Lubricant Analysis System, featuring customized workflows for instrument operation and diagnostics. The package includes HATR Setup workflows for background measurement, cell calibration, reference collection, and cleaning checks, and HATR In-Service Lubricant workflows for direct and subtractive petroleum oil analysis based on ASTM standards.
Use OMNIC Paradigm software Operator mode to operate the Lubricant Analysis System. The main window provides features for installing and running customized workflows to operate the instrument and instrument diagnostics.
There are two categories of workflows in this package:
  • HATR Setup workflows
  • HATR In-Service Lubricant workflows
HATR setup workflows
  • Run Background. Performs a background measurement with the instrument. The background should be measured every 2 hours during continuous use.
  • Calibrate Cell. Measures known material (squalane) to determine sample pathlength. Calculates pathlength and offset values used in sample measurements.
To ensure accurate analysis results, we recommend that you run the Calibrate Cell workflow at least once each day of instrument use and if you use a different HATR accessory or crystal.
  • Collect Reference. Measures a reference oil sample. Uses the stored reference for sample measurements taken with a “differential” sample workflow. 
  • Clean Check. Performs a sample measurement through the ATR crystal without a sample in place to determine if the windows have been properly cleaned.
HATR in-service lubricant workflows
  • ASTM E2412 Direct Petroleum. Intended for petroleum-based sample oils to be measured using the ASTM E2412 standard practice.
  • ASTM E2412 Subtract Petroleum. Intended for petroleum-based sample oils that have a matching pure reference oil, where the reference measurement is subtracted from the sample measurement. Use the Collect Reference workflow to measure and archive the reference oil. 
  • ASTM D7418 Direct Petroleum. Intended for any sample oil to be measured using the ASTM D7418 standard practice
  • ASTM D7418 Subtract Petroleum. Intended for petroleum-based sample oils that have a matching pure reference oil, where the reference measurement is subtracted from the sample measurement. Use the Collect Reference workflow to measure and archive the reference oil.
Symbol Meaning
hatricon1.png Categories for the HATR (Horizontal ATR) lubricant analysis system.
hatricon2.png Workflows that measure sample oil content directly (without a reference oil). Direct sample workflows can be based on the ASTM E2412 or D7418 method.
hatricon3.png Workflows that use a pure reference oil measurement to determine sample oil content.
  • Was this article helpful?