Thermo Fisher Scientific
269-3637 Revision A Nicolet Summit X HATR Quick Start Guide
This guide demonstrates how to set up the Thermo Scientific™ Affirma™ Horizontal ATR Lubricant Analysis System for the Nicolet™ Summit™ X Fourier transform infrared (FT-IR) spectrometer and perform a basic analysis.
The system consists of a Pike Horizontal Attenuated Total Reflection (HATR) accessory and baseplate that fit in the spectrometer sample compartment.
Our OMNIC Paradigm software and customized workflows step you through each sample analysis.
Figure 1-1: Nicolet Summit X HATR Lubricant Analysis System
Summit X Pike HATR.jpg
PDF downloads
Nicolet Summit X HATR Quick Start Guide (PDF) - Thermo Fisher Scientific
  • Was this article helpful?