Adding a macro to the OMNIC toolbar
To add a macro to the OMNIC toolbar, go to Edit: Edit Toolbar, select Add Item, choose Run Basic Macro, browse to the macro location, modify the item name and icon, drag the macro icon to the desired toolbar location, click OK, save the configuration, and click the icon to run the macro when needed.
Issue
How to add a macro file as on icon on the OMNIC toolbar
Environment
OMNIC
Resolution
The attached document has the steps for adding a macro to the OMNIC toolbar and then saving the settings so they are persistent.
Attachment(s)
| File | Last Modified |
|---|---|
| Add a macro to the OMNIC toolbar.pdf | February 06, 2023 |