Thermo Fisher Scientific
Error On Startup of Picta Software - Object Reference Not Set to an Instance of an Object
An error message may appear when opening OMNIC Picta due to a corrupted default experiment file, though the software continues to function normally. Replacing or clearing the corrupted file resolves the issue.
Issue
When opening the Picta software, an error message shows up that states "Object Reference Not Set to an Instance of an Object".  Clicking continue allows Picta to open normally and the software does not seem to have any problems.
Environment
Nicolet iN10
OMNIC Picta
Resolution
There are different ways to correct this problem.
  1. Open Picta and on the View and Collect menu, clear the default experiment so that no experiment files are loaded on startup.
  2. Delete the corrupted experiment file and create a new one from scratch.  Then use the new experiment file as the default to load on startup.
  3. If using the default_picta.picta file, delete the file from the C:\my documents\omnic\param\ folder and then copy the same file from C:\my documents\omnic\param\factory\ back into the Param folder.
Cause
This error is caused by a corrupted experiment file which has been set as the default to load on startup.  
A similar message can happed if the user tries to open a corrupted experiment file when Picta is already open.