Create and Customize Microscopy Reports
Create and customize microscopy reports
After analyzing a sample, you can print or save a report to record your findings.
❖ To create a microscopy report
- From the Session Analysis tab, go to File > Create Report.
- Enter a title for the report, select a format, and choose a template. The available templates depend on the type of analysis you performed.
- To customize the report, select Options and choose elements to include or exclude.
- Click Create.
Copy and export mosaic images
Right-click the mosaic image or mosaic sky view to copy or export the image as an image file. This is a good option when you want to add the image to a document or presentation rather than create a full report.
❖ To export a mosaic image
-
In a session analysis tab, right-click the mosaic image.
-
Select Export mosaic to save the image as a PNG, JPEG, or BMP file.
-
To print the mosaic, right-click and choose Print mosaic to send the image to the print preview and print it.
The image will be exported or printed as it is currently shown on the screen. To save or print the full image, ensure you are zoomed out to view the entire mosaic before right-clicking .