View Measurement History
View the measurement history to compare a spectrum at different processing stages. Navigate to the history panel in Spectral View, right-click an item, and select Measurement History. The historical data shows the spectrum at each processing stage. Create a report by selecting the Historical Spectrum template from the File menu. Exit by clicking Close Measurement History.
View the measurement history to visually compare a spectrum at different stages of processing and analysis. For example, after applying automatic baseline correction, view the measurement history to quickly see how the spectrum changed.
❖ To view the measurement history
- From the Spectral View, navigate to the history panel.
- Right-click any item in the history and select Measurement History.
The historical data displays the spectrum at each stage in your processing and analysis.
- To create a report of the historical data, open the File menu and select Create Report. Select the Historical Spectrum template, edit your report settings, and click Create.
- To exit the measurement history, click Close Measurement History.