Database File Location When Using SQLite for Audit Manager
Issue
What is the database file location when using SQLite for Audit Manager?
Environment
OMNIC Security Suite v2.1 software or newer
Resolution
When installing the OMNIC Security Suite v2.1 software or newer, the Audit Manager program is also installed. If doing a local installation and selecting the SQLite database option for the Audit Manager, the file used to store the audit log information has a file name of "AuditManager.DB" and is stored in the "C:\Program Data\Thermo Scientific\Audit Log\" directory.
The permission for this folder is initially set by the software and should be Full Access for the Audit Log Service Account, and Read-only (i.e. Read, List folder contents, and Read & Execute) for all other users.