Thermo Fisher Scientific
System failing the lamp test on startup.
Issue
Lamp Failing on startup
Environment
  • Spectronic 200e
Resolution
Lamp failed message most likely means the lamp has failed and needs to be replaced the replacement part number is below.
  1. Part number 335423-000 Spare lamps for SPEC200E and GENESYS 20, pkg of 2
  • Was this article helpful?