Thermo Fisher Scientific
VISIONlite Demo Software
Issue
Is there a demo version of the VISIONlite PC software?
Environment
VISIONlite is compatible with:
  • SPECTRONIC 200E
  • GENESYS 3S
  • GENESYS 10S series
  • GENESYS 20
  • BioMate 3S
  • Evolution 60S
These instruments can use VISIONlite 5 with a Serial to USB adapter:
  • Evolution 300
  • Helios series
Resolution
Download a demo version of the software from this website:
http://ascanis.com/Visitor_Downloads/visitor_downloads.html