Chromeleon is not recognizing license
Issue
Chromeleon is not recognizing my license
Environment
- Chromeleon 6
- Chromeleon 7
Resolution
Proceed with these steps only if you are a full Chromeleon customer.
Chromeleon 7
- Check to make sure your License Dongle is in the computer in a USB 2.0 (not labelled SS, or blue). Unplug and replug to make sure the computer reacts to recognizing the dongle.
- If it is not recognized in multiple ports of the computer, contact a technical support technician to issue you a replacement.
- Go to the Start Menu, Thermo Chromeleon 7 folder, Administration Console.
- Under License Manager, go to Update License File.
- Your License File should be located under C:\ProgramData\Dionex\Chromeleon\
- If you can’t locate your license file, contact a technical support technician with your dongle serial number and ask for the license file.
- Once received, save the license to the C:\ProgramData\Dionex\Chromeleon, and follow the steps above.
- Close the Administration Console.
- Restart your computer.
- Open Chromeleon.
- If you are still getting a license error, contact a technical support technician.
Chromeleon 6
- Check to make sure your License Dongle is in the computer in a USB 2.0 (not labelled SS, or blue). Unplug and replug to make sure the computer reacts to recognizing the dongle.
- If it is not recognized in multiple ports of the computer, contact a technical support technician to issue you a replacement.
- Open Chromeleon 6.8, Help, About Chromeleon Client, Preferences.
- Check and verify the Key Code is entered and correct, and you have selected DONGLE.
- If you do not have the key code, contact a technical support technician with your dongle serial number and ask for the license key code.
- Press OK. Close Chromeleon.
- Restart your computer.
- Open Chromeleon.
- If you are still getting a license error, contact a technical support technician.