Chromeleon no longer converts Mass Spec data to .cdf
Issue
Chromeleon no longer converts Mass Spectrometer data to .cdf.
Environment
Chromeleon
Resolution
Refer to the attached document to address this issue.
Analysis
Mass Spectrometer data is considered 3D data and Chromeleon only converts the .cmbx file to .cdf for 2D data. Because of this, customers request how to convert to .cdf.
Attachment(s)
File | Last Modified |
---|---|
Chromeleon Guide To - CDF Export.pdf | January 14, 2022 |