065448 Part F - Rev 7.30a - Chromeleon Operational Qualification and Performance Qualification Software Templates for ICS-5000, ICS-5000+, ICS-6000 Capillary Instruments - User's Guide
Description:
It is recommended that the most current revision of the OQ-PQ software templates is used in order for a qualification to be considered valid. Operational Qualification and Performance Qualification are system-specific test procedures.
Environment:
- Chromeleon version 7.1 or higher
Module Minimum Chromeleon Software Version
- AS-AP 7.1 SR1 DU0C
- AS 7.1 SR1
- AS-DV 7.1 SR1
- ICS-5000 SP or DP 7.1 SR1
- ICS-5000+ SP or DP 7.1 SR1
- ICS-6000 SP or DP 7.1 SR1
- ICS-5000 SP or DP 7.1 SR1
- ICS-5000+ SP or DP 7.1 SR1
- ICS-6000+ SP or DP 7.1 SR1
- ICS-5000 EG 7.1 SR1
- ICS-5000+ EG 7.1 SR1
- ICS-6000 EG 7.1 SR1
- ICS-5000 EG 7.1 SR1
- ICS-5000+ EG 7.1 SR1
- ICS-6000+ EG 7.1 SR1
- ICS-5000 DC 7.1 SR1
- ICS-5000+ DC 7.1 SR1
- ICS-6000 DC 7.1 SR1
- ICS-5000 DC 7.1 SR1
- ICS-5000+ DC 7.1 SR1
- ICS-6000+ DC 7.1 SR1
- ICS-5000 CD 7.1 SR1
- ICS-5000+ CD 7.1 SR1
- ICS-6000 CD 7.1 SR1
- ICS-5000 CD 7.1 SR1
- ICS-5000+ CD 7.1 SR1
- ICS-6000+ CD 7.1 SR1
- ICS-5000 ED 7.1 SR1
- ICS-5000+ ED 7.1 SR1
- ICS-6000 ED 7.1 SR1
- ICS-5000 ED 7.1 SR1
- ICS-5000+ ED 7.1 SR1
- ICS-6000+ ED 7.1 SR1
- ICS Series VWD 7.1 SR1
- FLUKE189-289 7.1 SR1
- Pump:
- ICS-5000 SP
- ICS-5000+ SP
- ICS-6000 SP
- ICS-5000 DP Capillary version
- ICS-5000+ DP Capillary version
- ICS-6000 DP Capillary version
- Manual injection or any of the following autosamplers:
- AS
- AS-DV
- AS-AP
- AS-AP autosampler:
- 10 mL tray PN 074938 with
- ICS-5000 CD push mode
- ICS-5000+ CD push mode
- ICS-6000 CD push mode
- 1.5 mL tray PN 074936 with
- ICS-5000 CD or ED push mode only
- ICS-5000+ CD or ED push mode only
- ICS-6000 CD CD or ED push mode only
- The optional conductivity/pH module for the AS-AP is currently not supported in OQ-PQ. . To qualify the AS-AP, a 250 μL injection syringe is required. The buffer size should always be set at 1200 µL.
- 10 mL tray PN 074938 with
- Conductivity detector:
- ICS-5000 CD Capillary version
- ICS-5000+ CD Capillary version
- ICS-6000 CD Capillary version
- Electrochemical detector:
- ICS-5000 ED Capillary version
- ICS-5000+ ED Capillary version
- ICS-6000 ED Capillary version
- Eluent Generator:
- ICS-5000 EG
- ICS-5000+ EG
- ICS-6000 EG