Tracefinder not setting LC and MS to standby
Issue
Tracefinder is not setting the LC and MS to standby ONLY when a whole batch or more than one sample is submitted but it will set the LC and MS to standby when only one sample is submitted.
Environment
- Mass Spectrometry
- TraceFinder
Resolution
Go to TraceFinder configuration and verify that Full sequence submission is selected in the Acquisition submission options.
Attachment(s)
| File | Last Modified |
|---|---|
| Tracefinder acquisition submission options.png | January 10, 2022 |