TraceFinder Not Processing Data File
Issue
The file status does not turn from yellow to green as displayed below. There is no error reported.
Environment
- TraceFinder
- Mass Spectrometry
Resolution
Check the method peak detection of the compounds to verify if any compound is using Range Integrated or Range Detected in conjuction with having the fragment confirming ion enabled.
If this is the case then you will need to change the detection type to Single Detected and save the method. Tracefinder will now be able to process the data file. This is a known issue and will be fixed in Tracefinder version 5.0.
Cause
If you encounter a problem with Tracefinder not processing some data files while other data files process ok there may be an issue with the method peak detection settings.
Attachment(s)
| File | Last Modified |
|---|---|
| not processing data file - image 02.png | January 10, 2022 |
| not processing data file - image 01.png | January 10, 2022 |