Thermo Fisher Scientific
FTIR Crystal Reference Guide
Issue
Determining the crystal materials best suited for infrared sampling needs
Environment
Fourier Transform Infrared systems
Resolution
Major considerations in choosing materials are:
  • Frequency range,
  • Solubility of the sample,
  • Refractice Index,
  • Reactivity with the sample, and
  • Mechanical and thermal characteristics.
Use the attached Crystal Reference Guide to determine the crylstal materials best suited for infrared sampling needs.
  • Was this article helpful?