Burner Optimization
Issue
How to optimize the position of the light patch in respect to the burner head
Environment
- iCE 3300
- iCE 3500
Resolution
The video below instructs how to utilize the different adjustments to maximize the position of the burner in respect to the light patch
NOTE: This video is demonstrated on a model of flame AA that did not have the door interlock sensor. If the instrument you have has the door interlock sensor, you will need to use the adjustment screw on the door to move the burner in and out of the light beam.
Analysis
It is a good idea to optimize the position of the burner head when experiencing issues such as low sensitivity