Burner ignitor adjustment
Issue
If the igniter arm is not in the correct position the flame will not light or the igniter may be in the optical path
Environment
- iCE3300
- iCE3500
- S Series
- M6
Resolution
See the video example of aligning the igniter. The written procedure is attached as well.
The written procedure is attached as well.
Attachment(s)
File | Last Modified |
---|---|
How to check the correct position of the burner igniter arm.pdf | August 02, 2022 |