Burner ignitor adjustment
The igniter arm can get bumped out of position causing the system to not ignite the flame. Use the supplied burner slot cleaning/igniter arm alignment tool to check the correct position of the arm. The tool is in a slot on the right side of the burner base. It is removed by sliding the tool forward from the base. This article provides a video and a written procedure for repositioning the arm. This article applies to the iCE3300/3500 instruments.
Issue
If the igniter arm is not in the correct position the flame will not light or the igniter may be in the optical path
Environment
- iCE3300
- iCE3500
- S Series
- M6
Resolution
See the video example of aligning the igniter. The written procedure is attached as well.
The written procedure is attached as well.
Attachment(s)
| File | Last Modified |
|---|---|
| How to check the correct position of the burner igniter arm.pdf | August 02, 2022 |