Thermo Fisher Scientific
Flame issue
Issue
Flame is yellow and white at the base of the flame
Environment
  • iCE 3300
  • iCE 3500
Resolution
Perform Setup Flame
Analysis
The ratio of oxidant to fuel is off, setup flame adjusts this ratio.
  • Was this article helpful?