Monochromator Calibration
Issue
Sometimes it is necessary to perform a monochromator calibration. This could be for example if the wavelength is not matching up exactly with the one specified in the method. Or, If error messages such as MD2019, MD2066, MD2070 are encountered when setting up the Optics and the light patch is observed around the entrance window.
Environment
- iCE 3000 Series
Resolution
1) Insert Ca/Mg lamp in carousel position 6
2) Disconnect communications in SOLAAR
3) Open OQTest Software
4) Connect in OQTest software
5) Click on Calibrate button