Thermo Fisher Scientific
Flame issue
In the iCE 3300 and iCE 3500 environments, the flame may appear yellow and white at its base. This issue is due to an incorrect oxidant-to-fuel ratio. To resolve this, perform the Setup Flame procedure, which adjusts the ratio to the correct levels, ensuring proper flame characteristics for accurate analysis.
Issue
Flame is yellow and white at the base of the flame
Environment
  • iCE 3300
  • iCE 3500
Resolution
Perform Setup Flame
Analysis
The ratio of oxidant to fuel is off, setup flame adjusts this ratio.
  • Was this article helpful?