Boron Carryover in iCAP
Issue
Long time needed for Boron to rinse out of sample introduction system
Environment
- 61E
- iCAP 6500 Radial
- iCAP 6500 Dual
- iCAP 6300 Radial
- iCAP 6200 Dual
- iCAP 6300 Dual
- INTREPID
- IRIS
- TRACE
Resolution
- Add Triton X-100 to the rinse solution (0.1%). It has been found that this has helped along with increasing the rinse time.
- Try decreasing the Boron concentration the next time calibration standards are made.