Blue Indicator Light for Detector Temperature in Qtegra
Issue
Blue indicator light for Detector Temperature in Qtegra
Environment
- iCAP 6000 Series
- iCAP 7000 Series
Resolution
The blue icon in Qtegra Dashboard is only an indication that the detector temperature is lower than -50°C which is not a problem for measurements. The iCAP will still perform perfectly well and no immediate action needs to be taken.
However, if one sees drifting peak positions and this temperature reads -55 degrees ( in iTEVA), then the Camera needs replacing.
Please note: The chiller needs to be switched on for the camera to cool.
To check the exact temperature in iTEVA, perform the following:
1) Open iTEVA
2) Select admin from dropdown menu and click Ok
3) Click the Instrument Status icon
4) The detector (Camera) Temperature will be displayed in this window.
Attachment(s)
File | Last Modified |
---|---|
Camera Temperature in Instrument Status.jpg | September 13, 2022 |
Instrument Status Icon.jpg | September 13, 2022 |
iTEVA Login.jpg | September 13, 2022 |
iTEVA icon.jpg | September 13, 2022 |