Disabling / Enabling the Drain Sensor in Qtegra
Issue
Disabling the Drain Sensor in Qtegra
Environment
- iCAP 6000
- iCAP 7000
- Qtegra
Resolution
- You must first close Qtegra
- Then open Configurator
- Select Experiment Configurator
- Click the arrow next to the configuration of interest to reveal components
- Then right click EDIT to see SETTINGS
- Under iCAP OES, find "Drain Flow Sensor Enabled" and click the drop down
- Select False to turn drain sensor off or True to enable it
- After you change it, hit the Save icon and close Configurator.
- You can then reopen Qtegra
Cause
Customer or application determines that the drain sensor is a hindrance and needs to be turned off.
- Capillary type does not allow light passage for sensor to function
- Sample viscosity prevents sensor from functioning
- Customer elects to turn off drain sensor
Attachment(s)
File | Last Modified |
---|---|
Configurator icon (002).png | August 02, 2022 |
Experiment Configurator icon (002).png | August 02, 2022 |
clipboard_e840d9b65b5fb13c2681abdf6e8254cb6.png | August 02, 2022 |
TRUE_FALSE.jpg | August 02, 2022 |
New Save Delete Load.jpg | August 02, 2022 |