Thermo Fisher Scientific
High Internal Standard Recovery on iCAP 6000-7000 series
Issue
High Internal Standard Recovery 
Environment
  • iCAP 6000 Series
  • iCAP 7000 Series
Resolution
Somehow purge flow got set to High. Once it was set back to Normal, the Internal Standard Recoveries came back down near 100%
Analysis
Internal Standard Recoveries are going to ~125% during calibration and stay there afterwards. Internal standard was doing its job as QC recoveries were within tolerance. Customer was concerned thought that Internal Standard recoveries were 125% This was a wear metal analysis application.