Thermo Fisher Scientific
Humidity in the camera is too high
Issue
Detector is failing to cool. An error is being displayed that indicates that the humidity in the camera is too high.
Environment
  • iCAP PRO
Resolution
Enable the Engineering Purge
Analysis
The Engineering purge allows a higher level of gas flow through the Detector in order to potentially outgas any moisture present