Nebulizer/Spray Chamber Connector Kit
Description:
The Sample Introduction Tubing Spares Kit has been revised to include more drain and nebulizer PTFE fitting w/capillary fittings and to also make just those tubings available separately.
P/N 842312051402 Nebulizer/Spray Chamber Connector Kit contains these:
- Adapter PTFE Blk Ring w/integral capillary (chamber drain), 6 EA
- Adapter PTFE Brn Ring w/integral capillary (neb uptake), 6 EA
- Fitting Neb Gas 4mm inline QC (Ezylok), 3 EA
Typical Sample Introduction Tubing Spares:
842312051751 Sample Intro Tubing and Connector Kit
The Kit contains new drain and nebulizer PTFE fitting with capillary, nebulizer gas tubing with check Valve, Nebulizer gas fitting (4mm push/fit connector), capillary tubing for original drain and sample inlet, Sipper Probes and Sampling O-ring kit.
Environment:
iCAP 6000 Series
Attachment(s):
File | Last Modified |
---|---|
430116622812 - Spray Chamber Connector Kit.pdf | August 02, 2022 |