Fitting the molecular sieve drying trap
Issue
How do I fit the molecular sieve drying trap?
Environment
- iCAP 6500 Radial
- iCAP 6500 Dual
- iCAP 6300 Radial
- iCAP 6200 Dual
- iCAP 6300 Dual
Resolution
The molecular sieve dying trap is mounted in the support on the side cover of the RF generator.
- Secure it in place with 2 ty-raps.
- Connect the camera purge gas from the gasbox to the inlet at the bottom.
- Connect the outlet at the top to the camera.
Note – The orientation is not important when the trap is first installed. However, if it is removed for any reason, it must be replaced in the same orientation.