Rev 3a - 480221 - Alignment Guide for ASX-560 and ASX-280 Autosamplers
Description:
When to Align the Autosampler
Alignment is critical to the autosampler’s ability to operate reliably.
Alignment should also be performed if you suspect that shipping or rough handling may have disturbed the autosampler’s chassis, or when an autosampler component such as the Z-drive has been replaced.
Origin Alignment
Origin alignment sets the location of the home position so that the sample probe is centered in the rinse station. This alignment is done by setting offsets in the autosampler firmware. No mechanical adjustments are required.
Full Alignment
Full alignment sets the location of the sample vials and standards vials in relation to the rinse station. This alignment is done by setting offsets in the autosampler firmware. No mechanical adjustments are required.
Use this guide to to perform an alignment when using versions of the ASX Dashboard software previous to 1.4.1: 480221 ASX-560 Alignment Guide 2019-07-03.pdf
If you are using a ASX Dashboard version 1.4.1 or later, utilize this alignment guide: Rev 4 - 480221 - Alignment Guide for ASX-280 and ASX-560 Autosamplers
Environment:
- ASX-280
- ASX-560
Attachment(s):
File | Last Modified |
---|---|
480221 ASX-560 Alignment Guide 2019-07-03.pdf | January 04, 2023 |