Setting Uptake Time for ESI Autosampler in Qtegra
Issue
Setting Uptake Time for ESI Autosampler in Qtegra
Environment
- Qtegra
- ESI SC 2/4/8/16DX
Resolution
- Open new lab book
- Go to ESI Autosampler, and choose Method Settings
- Select Settings in the Method Settings window and choose Analysis
- Under Sample adjust Uptake Time as needed
- Save and run lab book