Custom Autosampler Racks not showing up in iTEVA
Issue
Customer autosampler racks are not showing up in order to be selected when creatign an autosampler sequence in iTEVA
Environment
- iTEVA
Resolution
For Windows 7: Copy the files below into the C:\Program Files (x86)\Thermo\iTEVA\SysData.
For Windows XP: Copy the files below into the C:\Program Files\Thermo\iTEVA\SysData.
NOTE: Rename the original *.xml files with a suffix such as “org” to indicate they were the original files installed with the software.
- AutoSamplerConfigurations.xml
- AutoSamplerConfigurationSets.xml
- AutoSamplerRacks.xml
- MAP files (these will be named something like "CETAC 5x12 Enviro 5 Map.xls")
Note: If you need assistance with creating a customer autosampler layout, please contact our technical support team.