Thermo Fisher Scientific
FID module heater failure error
To troubleshoot a FID module heater failure on a Trace 1300, first try power cycling the GC. If the issue persists, replace the FID module's heater and PT100 RTD sensor. The spare parts are available as follows: PN 23044000 (Assy HTR NPD/FID TRC1300) and PN 23044026 (Assy PT100 NPD/FID TRC1300). While the replacement is not complicated, it is recommended that a Field Service Engineer perform the repair.
Issue
How do I troubleshoot a FID module heater failure?
Environment
Trace 1300
Resolution
  1. Try power cycling the Trace 1300 Series GC
  2. Replace the FID Module's Heater and PT100 RTD sensor. They are available as spare parts below:
  • PN 23044000 Assy HTR NPD/FID TRC1300
  • PN 23044026 Assy PT100 NPD/FID TRC1300
NOTE: The replacement is not complicated but it is suggested that a Field Service Engineer perform the repair.
  • Was this article helpful?